missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Pregnen-17alpha-ol-3,20-dione-2,2,4,6,6,21,21,21-d8 Hexanoate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Hydroxyprogesterone Caproate-2,2,4,6,6,21,21,21-D8 |
| Molecular Formula | C27 D8 H32 O4 |
| Quantity | 0.005 g |
| Synonym | 4-Pregnen-17α-ol-3,20-dione-2,2,4,6,6,21,21,21-d8 Hexanoate, Hydroxyprogesterone Caproate-d8, Hydroxyprogesterone Caproate-2,2,4,6,6,21,21,21-D8, 17α-Hydroxyprogesterone-2,2,4,6,6,21,21,21-d8 hexanoate |
| InChI Formula | InChI=1 S/C27H40O4/c1-5-6-7-8-24(30)31-27(18(2)28)16-13-23-21-10-9-19-17-20(29)11-14-25(19,3)22(21)12-15-26(23,27)4/h17,21-23 H,5-16H2,1-4H3/t21-,22+,23+,25+,26+,27+/m1/s1/i2D3,9D2,11D2,17 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@@H]3 [C@H](CC[C@@]4(C)[C@H]3 CC[C@]4(OC(=O)CCCCC)C(=O)C([2 H])([2 H])[2 H])[C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 R)-2,2,4,6,6-pentadeuterio-10,13-dimethyl-3-oxo-17-(2,2,2-trideuterioacetyl)-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-17-yl] hexanoate |
| Molecular Weight (g/mol) | 436.6534 |
| Formula Weight | 436.3429 g/mol |
| Percent Purity | 96 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction