Learn More
5-Hydroxy-L-tryptophan-4,6,7-d3 H2O, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy-L-tryptophan-4,6,7-d3 |
| CAS | 1276197-29-5 |
| Molecular Formula | C11 D3 H9 N2 O3 |
| Quantity | 0.01 g |
| Synonym | Oxitriptan-d3, 5-Hydroxy-L-tryptophan-d3, 5-Hydroxy-L-tryptophan-4,6,7-D3, (2 S)-2-Amino-3-(5-hydroxy-1 H-indol-3-yl-4,6,7-d3)propanoic acid |
| InChI Formula | InChI=1 S/C11H12N2O3/c12-9(11(15)16)3-6-5-13-10-2-1-7(14)4-8(6)10/h1-2,4-5,9,13-14 H,3,12H2,(H,15,16)/t9-/m0/s1/i1D,2 D,4 D |
| SMILES | [2 H]c1c([2 H])c2[nH]cc(C[C@H](N)C(=O)O)c2c([2 H])c1O |
| IUPAC Name | (2 S)-2-amino-3-(4,6,7-trideuterio-5-hydroxy-1 H-indol-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 223.243 |
| Formula Weight | 223.104 g/mol |
| Show More |