missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5b-Pregnan-3a,11b,21-triol-20-one-17a,21,21-d3, CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 3 alpha,11 beta,21-Trihydroxy-5 beta-pregnan-20-one-17,21,21-D3 |
| Molecular Formula | C21 D3 H31 O4 |
| Quantity | 0.005 g |
| Synonym | 5 b-Pregnan-3 A,11 b,21-triol-20-one-17 A,21,21-d3, 3α,11β,21-Trihydroxy-5β-pregnan-20-one-17,21,21-D3, 3α,5β-Tetrahydrocorticosterone-17,21,21-d3, Tetrahydrocorticosterone-d3 |
| InChI Formula | InChI=1 S/C21H34O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h12-17,19,22-24 H,3-11H2,1-2H3/t12-,13-,14+,15+,16-,17+,19-,20+,21+/m1/s1/i11D2,16 D |
| SMILES | [2 H]C([2 H])(O)C(=O)[C@@]1([2 H])CC[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@H]3 [C@@H](O)C[C@]12 C |
| IUPAC Name | 2,2-dideuterio-1-[(3 R,5 R,8 S,9 S,10 S,11 S,13 S,14 S,17 S)-17-deuterio-3,11-dihydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,9,11,12,14,15,16-tetradecahydrocyclopenta[a]phenanthren-17-yl]-2-hydroxyethanone |
| Molecular Weight (g/mol) | 353.5108 |
| Formula Weight | 353.2645 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction