Learn More
Amitriptyline-d6 HCl (N,N-dimethyl-d6), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Amitriptyline-d6 Hydrochloride |
| CAS | 203645-63-0 |
| Molecular Formula | C20 2H6 H17 N . Cl H |
| Quantity | 0.01 g |
| Synonym | 1-Propanamine, 3-(10,11-dihydro-5 H-dibenzo[a,d]cyclohepten-5-ylidene)-N,N-di(methyl-d3)-, hydrochloride (9 CI), Amitriptyline-d6 hydrochloride, Amitriptyline-D6 hydrochloride (N,N-dimethyl D6) |
| InChI Formula | InChI=1 S/C20H23N.ClH/c1-21(2)15-7-12-20-18-10-5-3-8-16(18)13-14-17-9-4-6-11-19(17)20;/h3-6,8-12 H,7,13-15H2,1-2H3;1 H/i1D3,2D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(CCC=C1c2ccccc2CCc3ccccc13)C([2 H])([2 H])[2 H] |
| IUPAC Name | 3-(5,6-dihydrodibenzo[1,2-a:2',1'-d][7 ]annulen-11-ylidene)-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride |
| Molecular Weight (g/mol) | 319.9 |
| Formula Weight | 319.1974 g/mol |
| Show More |