missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Beflubutamid-d7 (benzylamine-d7), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Beflubutamid D7 (Benzyl D7) |
| Molecular Formula | C18 D7 H10 F4 N O2 |
| Quantity | 0.005 g |
| Synonym | Beflubutamid D7 (Benzyl D7), 2-[4-Fluoro-3-(trifluoromethyl)phenoxy]-N-[(phenyl-d5)methyl-d2]butanamide, Beflubutamid-d7, (±)-Beflubutamid-d7 (benzylamine-d7), Benfluamid-d7, N-(Benzyl-d7)-2-(4-fluoro-3-(trifluoromethyl)phenoxy)butanamide |
| InChI Formula | InChI=1 S/C18H17F4NO2/c1-2-16(17(24)23-11-12-6-4-3-5-7-12)25-13-8-9-15(19)14(10-13)18(20,21)22/h3-10,16 H,2,11H2,1H3,(H,23,24)/i3D,4 D,5 D,6 D,7 D,11D2 |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])NC(=O)C(CC)Oc2ccc(F)c(c2)C(F)(F)F |
| IUPAC Name | N-[dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl]-2-[4-fluoro-3-(trifluoromethyl)phenoxy]butanamide |
| Molecular Weight (g/mol) | 362.3698 |
| Formula Weight | 362.1635 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction