missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bisphenol S-2,2',3,3',5,5',6,6'-d8, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Bisphenol S (ring-D8) |
| CAS | 2483831-28-1 |
| Molecular Formula | C12 D8 H2 O4 S |
| Quantity | 0.01 g |
| Synonym | Phen-2,3,5,6-d4-ol, 4,4'-sulfonylbis- (ACI): 4,4'-Sulfonylbis[phen-2,3,5,6-d4-ol] (ACI), 4,4'-Sulfonylbis[phenol-2,3,5,6-d4], 1,1'-Sulfonylbis[4-hydroxybenzene-2,3,5,6-d4], 4,4'-Bisphenol S (ring-D8), 4,4'-Dihydroxydiphenyl sulfone (2,2',3,3',5,5',6,6'-D8), p,p'-Dihydroxydiphenyl sulfone (ring-D8), p,p'-Dihydroxydiphenyl sulfone (ring-D8), p,p'-Dihydroxydiphenyl sulfone (ring-D8), p,p'-Dihydroxydiphenyl sulfone (ring-D8) |
| InChI Formula | InChI=1 S/C12H10O4S/c13-9-1-5-11(6-2-9)17(15,16)12-7-3-10(14)4-8-12/h1-8,13-14 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c(c([2 H])c([2 H])c1O)S(=O)(=O)c2c([2 H])c([2 H])c(O)c([2 H])c2[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-4-(2,3,5,6-tetradeuterio-4-hydroxyphenyl)sulfonylphenol |
| Molecular Weight (g/mol) | 258.3197 |
| Formula Weight | 258.0802 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction