Learn More
Cholesterol-25,26,26,26-d4, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Cholesterol 6,7,7-d4 |
| CAS | 956029-28-0 |
| Molecular Formula | C27 2H4 H42 O |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28 H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i1D3,18 D/t18?,19-,21+,22+,23-,24+,25+,26+,27- |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C)CCC[C@@H](C)[C@H]1 CC[C@H]2 [C@@H]3 CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3 CC[C@]12 C |
| IUPAC Name | (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6,7,7,7-tetradeuterio-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol |
| Molecular Weight (g/mol) | 390.68 |
| Formula Weight | 390.38 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |