Learn More
(+)-cis-Diltiazem-d3 HCl (acetoxy-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (+)-cis-Diltiazem-d3 HCl (acetoxy-d3) |
| CAS | 1217860-13-3 |
| Molecular Formula | C22 D3 H23 N2 O4 S . Cl H |
| Quantity | 0.01 g |
| Synonym | (2 S,3 S)-3-(Acetyloxy-d3)-5-[2-(dimethylamino)ethyl]-2,3-dihydro-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5 H)-one hydrochloride (1:1), Diltiazem-d3 hydrochloride, (+)-cis-Diltiazem-d3 hydrochloride, Diltiazem-D3 (acetyl-D3) hydrochloride, (+)-cis-Diltiazem-d3 HCl (acetoxy-d3) |
| InChI Formula | InChI=1 S/C22H26N2O4S.ClH/c1-15(25)28-20-21(16-9-11-17(27-4)12-10-16)29-19-8-6-5-7-18(19)24(22(20)26)14-13-23(2)3;/h5-12,20-21 H,13-14H2,1-4H3;1 H/t20-,21+;/m1./s1/i1D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C(=O)O[C@@H]1 [C@@H](Sc2ccccc2N(CCN(C)C)C1=O)c3ccc(OC)cc3 |
| IUPAC Name | [(2 S,3 S)-5-[2-(dimethylamino)ethyl]-2-(4-methoxyphenyl)-4-oxo-2,3-dihydro-1,5-benzothiazepin-3-yl] 2,2,2-trideuterioacetate;hydrochloride |
| Molecular Weight (g/mol) | 453.9973 |
| Formula Weight | 453.1568 g/mol |
| Show More |