missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dehydroepiandrosterone-2,2,3,4,4,6-d6, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Dehydroepiandrosterone (DHEA) (2,2,3,4,4,6-D6) |
| CAS | 1261254-39-0 |
| Molecular Formula | C19 D6 H22 O2 |
| Quantity | 0.005 g |
| InChI Formula | InChI=1 S/C19H28O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h3,13-16,20 H,4-11H2,1-2H3/t13?,14?,15?,16?,18-,19-/m0/s1/i3D,7D2,11D2,13 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C([2 H])(O)C([2 H])([2 H])C[C@]2(C)C3CC[C@@]4(C)C(CCC4=O)C3C1 |
| IUPAC Name | (10 R,13 S)-2,2,3,4,4,6-hexadeuterio-3-hydroxy-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-1 H-cyclopenta[a]phenanthren-17-one |
| Molecular Weight (g/mol) | 294.46 |
| Formula Weight | 294.2466 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction