missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desloratadine-d4 (piperidinylidene-3,3,5,5-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Desloratadine-D4 (piperidine-3,3,5,5-D4) |
| CAS | 2713301-38-1 |
| Molecular Formula | C19 D4 H15 Cl N2 |
| Quantity | 0.01 g |
| Synonym | 8-Chloro-6,11-dihydro-11-(4-piperidinylidene-3,3,5,5-d4)-5 H-benzo[5,6 ]cyclohepta[1,2-b]pyridine, Desloratadine-d4, Desloratadine-D4 (piperidine-3,3,5,5-D4), Desloratadine-d4 (piperidinylidene-3,3,5,5-d4) |
| InChI Formula | InChI=1 S/C19H19ClN2/c20-16-5-6-17-15(12-16)4-3-14-2-1-9-22-19(14)18(17)13-7-10-21-11-8-13/h1-2,5-6,9,12,21 H,3-4,7-8,10-11H2/i7D2,8D2 |
| SMILES | [2 H]C1([2 H])CNCC([2 H])([2 H])C1=C2c3ccc(Cl)cc3CCc4cccnc24 |
| IUPAC Name | 13-chloro-2-(3,3,5,5-tetradeuteriopiperidin-4-ylidene)-4-azatricyclo[9.4.0.03,8 ]pentadeca-1(11),3(8),4,6,12,14-hexaene |
| Molecular Weight (g/mol) | 314.8452 |
| Formula Weight | 314.1488 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction