missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diflufenican-d3 (3-trifluoromethylphenoxy-2,4,6-d3), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Diflufenican D3 (3-trifluoromethylphenoxy-2,4,6 D3) |
| CAS | 1185009-29-3 |
| Molecular Formula | C19 2H3 H8 F5 N2 O2 |
| Quantity | 0.01 g |
| Synonym | Diflufenican D3 (3-trifluoromethylphenoxy-2,4,6 D3) |
| InChI Formula | InChI=1 S/C19H11F5N2O2/c20-12-6-7-16(15(21)10-12)26-17(27)14-5-2-8-25-18(14)28-13-4-1-3-11(9-13)19(22,23)24/h1-10 H,(H,26,27)/i3D,4 D,9 D |
| SMILES | [2 H]c1cc([2 H])c(c([2 H])c1Oc2ncccc2C(=O)Nc3ccc(F)cc3F)C(F)(F)F |
| IUPAC Name | N-(2,4-difluorophenyl)-2-[2,4,6-trideuterio-3-(trifluoromethyl)phenoxy]pyridine-3-carboxamide |
| Molecular Weight (g/mol) | 397.31 |
| Formula Weight | 397.0929 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction