Learn More
Indeno[1,2,3-c,d]pyrene-d12, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Indeno(1,2,3-c,d)pyrene D12 |
| CAS | 203578-33-0 |
| Molecular Formula | C22 2H12 |
| Quantity | 0.005 g |
| Synonym | Indeno[1,2,3-cd]pyrene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Indeno[1,2,3-cd]pyrene-d12 (9 CI), Indeno[1,2,3-c,d]pyrene D12, Indeno(1,2,3-c,d)pyrene D12 |
| InChI Formula | InChI=1 S/C22H12/c1-2-7-17-16(6-1)18-11-10-14-9-8-13-4-3-5-15-12-19(17)22(18)21(14)20(13)15/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c4c([2 H])c([2 H])c5c([2 H])c([2 H])c([2 H])c6c([2 H])c2c3c4c56 |
| IUPAC Name | Indeno[1,2,3-cd]pyrene-1,2,3,4,5,6,7,8,9,10,11,12-d12 |
| Molecular Weight (g/mol) | 288.4 |
| Formula Weight | 288.1692 g/mol |
| Show More |