Learn More
4-Chlorobenzoic-d4 Acid, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 4-Chlorobenzoic acid ring-d4 |
| CAS | 85577-25-9 |
| Molecular Formula | C7 2H4 H Cl O2 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C7H5ClO2/c8-6-3-1-5(2-4-6)7(9)10/h1-4 H,(H,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(C(=O)O)c([2 H])c([2 H])c1Cl |
| IUPAC Name | 4-chloro-2,3,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 160.59 |
| Formula Weight | 160.0229 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |