missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Kynurenine-d4 [4-(2-aminophenyl-3,5-d2); 3,3-d2], CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | L-Kynurenine-d4 [4-(2-aminophenyl-3,5-d2); 3,3-d2] |
| CAS | 2672568-86-2 |
| Molecular Formula | C10 D4 H8 N2 O3 |
| Quantity | 0.01 g |
| Synonym | Benzene-3,5-d2-butanoic-β,β-d2 acid, α,2-diamino-γ-oxo-, (αS)- (ACI), (αS)-α,6-Diamino-γ-oxobenzene-3,5-d2-butanoic-β,β-d2 acid, (αS)-α,2-Diamino-γ-oxobenzenebutanoic acid-d4, (S)-3-Anthraniloylalanine-d4, L-Kynurenine-d4, (2 S)-2-Amino-4-(2-aminophenyl-3,5-d2)-4-oxobutanoic-3,3-d2 acid, (2 S)-2-Amino-4-(2-aminophenyl-3,5-d2)-4-oxobutanoic-3,3-d2 acid, (2 S)-2-Amino-4-(2-aminophenyl-3,5-d2)-4-oxobutanoic-3,3-d2 acid |
| InChI Formula | InChI=1 S/C10H12N2O3/c11-7-4-2-1-3-6(7)9(13)5-8(12)10(14)15/h1-4,8 H,5,11-12H2,(H,14,15)/t8-/m0/s1/i1D,4 D,5D2 |
| SMILES | [2 H]c1cc([2 H])c(N)c(c1)C(=O)C([2 H])([2 H])[C@H](N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-4-(2-amino-3,5-dideuteriophenyl)-3,3-dideuterio-4-oxobutanoic acid |
| Molecular Weight (g/mol) | 212.2385 |
| Formula Weight | 212.1099 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction