Learn More
L-Tryptophan-2,3,3-d3, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | L-Tryptophan-d3 |
| CAS | 133519-78-5 |
| Molecular Formula | C11 D3 H9 N2 O2 |
| Quantity | 0.01 g |
| Synonym | L-Tryptophan-α,β,β-d3 (9 CI), α,β,β-Tri-2 H-L-tryptophan, L-Tryptophan-α,β,β-d3, L-Tryptophan-d3, (S)-α-Amino-3-indole-α,β,β-d3-propionic acid, (αS)-α-Aminoindole-3-propanoic-d3 acid, L-Tryptophan-α,β,β-D3 |
| InChI Formula | InChI=1 S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13 H,5,12H2,(H,14,15)/t9-/m0/s1/i5D2,9 D |
| SMILES | [2 H]C([2 H])(c1c[nH]c2ccccc12)[C@]([2 H])(N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-2,3,3-trideuterio-3-(1 H-indol-3-yl)propanoic acid |
| Molecular Weight (g/mol) | 207.244 |
| Formula Weight | 207.109 g/mol |
| Show More |