Learn More
(±)-Metoprolol-d7 HCl (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Metoprolol-D7 Hydrochloride |
| CAS | 1219798-61-4 |
| Molecular Formula | C15 D7 H18 N O3 . Cl H |
| Quantity | 0.01 g |
| Synonym | 1-[4-(2-Methoxyethyl)phenoxy]-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]-2-propanol hydrochloride (1:1), 1-Isopropyl-d7-amino-3-[4-(2-methoxyethyl)phenoxy]-2-propanol hydrochloride, (±)-Metoprolol-d7 HCl (iso-propyl-d7), Metoprolol-d7 Hydrochloride, Metoprolol-D7 (N-isopropyl-D7) Hydrochloride |
| InChI Formula | InChI=1 S/C15H25NO3.ClH/c1-12(2)16-10-14(17)11-19-15-6-4-13(5-7-15)8-9-18-3;/h4-7,12,14,16-17 H,8-11H2,1-3H3;1 H/i1D3,2D3,12 D; |
| SMILES | Cl.[2 H]C([2 H])([2 H])C([2 H])(NCC(O)COc1ccc(CCOC)cc1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-3-[4-(2-methoxyethyl)phenoxy]propan-2-ol;hydrochloride |
| Molecular Weight (g/mol) | 310.868 |
| Formula Weight | 310.204 g/mol |
| Show More |