missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nalpha-Acetyl-L-ornithine-5,5-d2, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Nalpha-Acetyl-L-ornithine-d2 |
| Molecular Formula | C7 D2 H12 N2 O3 |
| Quantity | 0.01 g |
| Synonym | N2-Acetyl-L-ornithine-5,5-D2, (2 S)-5-Amino-2-acetamidopentanoic-5,5-d2 acid, Nα-Acetyl-L-ornithine-5,5-d2 |
| InChI Formula | InChI=1 S/C7H14N2O3/c1-5(10)9-6(7(11)12)3-2-4-8/h6H,2-4,8H2,1H3,(H,9,10)(H,11,12)/t6-/m0/s1/i4D2 |
| SMILES | [2 H]C([2 H])(N)CC[C@H](NC(=O)C)C(=O)O |
| IUPAC Name | (2 S)-2-acetamido-5-amino-5,5-dideuteriopentanoic acid |
| Molecular Weight (g/mol) | 176.21 |
| Formula Weight | 176.113 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction