missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentamidine-d4 2HCl (pentane-1,1,5,5-d4), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Pentamidine-D4 (pentane-1,1,5,5-D4) Dihydrochloride |
| CAS | 1276197-32-0 |
| Molecular Formula | C19 D4 H20 N4 O2 . 2 Cl H |
| Quantity | 0.01 g |
| Synonym | 4,4'-[1,5-Pentane-1,1,5,5-d4-diylbis(oxy)]bisbenzenecarboximidamide dihydrochloride, 4,4'-(Pentamethylenedioxy-α,ω-d4)dibenzamidine dihydrochloride, Pentamidine-d4 dihydrochloride, Pentamidine-d4 2 HCl (pentane-1,1,5,5-d4), Pentamidine-D4 (pentane-1,1,5,5-D4) dihydrochloride |
| InChI Formula | InChI=1 S/C19H24N4O2.2 ClH/c20-18(21)14-4-8-16(9-5-14)24-12-2-1-3-13-25-17-10-6-15(7-11-17)19(22)23;;/h4-11 H,1-3,12-13H2,(H3,20,21)(H3,22,23);2*1 H/i12D2,13D2;; |
| SMILES | Cl.Cl.[2 H]C([2 H])(CCCC([2 H])([2 H])Oc1ccc(cc1)C(=N)N)Oc2ccc(cc2)C(=N)N |
| IUPAC Name | 4-[5-(4-carbamimidoylphenoxy)-1,1,5,5-tetradeuteriopentoxy]benzenecarboximidamide;dihydrochloride |
| Molecular Weight (g/mol) | 417.366 |
| Formula Weight | 416.168 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction