missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Permethrin-d9 (3-phenoxy-d5-benzyl-2,4,5,6-d4) (cis/trans mixture), CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Permethrin-d9 (3-phenoxy-d5-benzyl-2,4,5,6-d4) (cis/trans mixture) |
| Molecular Formula | C21 D9 H11 Cl2 O3 |
| Quantity | 0.01 g |
| Synonym | Permethrin D9 (Phenoxyphenyl D9), Permethrin D9 (3-phenoxyphenyl D9) |
| InChI Formula | InChI=1 S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19 H,13H2,1-2H3/i3D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(Oc2c([2 H])c([2 H])c([2 H])c(COC(=O)C3C(C=C(Cl)Cl)C3(C)C)c2[2 H])c([2 H])c1[2 H] |
| IUPAC Name | [2,3,4,6-tetradeuterio-5-(2,3,4,5,6-pentadeuteriophenoxy)phenyl]methyl 3-(2,2-dichloroethenyl)-2,2-dimethylcyclopropane-1-carboxylate |
| Molecular Weight (g/mol) | 400.343 |
| Formula Weight | 399.135 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction