Learn More
Phosmet-d6 (dimethyl-d6), CDN
Product Code. 30314529
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30314529
Brand: CDN CDND80650.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Phosmet D6 (dimethoxy D3)
CAS 2083623-41-8
Molecular Formula C11 2H6 H6 N O4 P S2
Quantity 0.01 g
Synonym Phosmet D6 (dimethyl D3)
InChI Formula InChI=1 S/C11H12NO4PS2/c1-15-17(18,16-2)19-7-12-10(13)8-5-3-4-6-9(8)11(12)14/h3-6 H,7H2,1-2H3/i1D3,2D3
SMILES [2 H]C([2 H])([2 H])OP(=S)(OC([2 H])([2 H])[2 H])SCN1C(=O)c2ccccc2C1=O
IUPAC Name 2-[bis(trideuteriomethoxy)phosphinothioylsulfanylmethyl]isoindole-1,3-dione
Molecular Weight (g/mol) 323.36
Formula Weight 323.0322 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity