Learn More
Quinolinic-4,5,6-d3 acid, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 2,3-Pyridinedicarboxylic Acid-d3 |
| CAS | 138946-42-6 |
| Molecular Formula | C7 D3 H2 N O4 |
| Quantity | 0.005 g |
| Synonym | 2,3-Pyridine-4,5,6-d3-dicarboxylic acid (9 CI), 2,3-Pyridinedicarboxylic Acid-4,5,6-D3, Quinolinic-4,5,6-d3 acid, Quinolinic acid-d3, 2,3-Pyridinedicarboxylic Acid-d3 |
| InChI Formula | InChI=1 S/C7H5NO4/c9-6(10)4-2-1-3-8-5(4)7(11)12/h1-3 H,(H,9,10)(H,11,12)/i1D,2 D,3 D |
| SMILES | [2 H]c1nc(C(=O)O)c(C(=O)O)c([2 H])c1[2 H] |
| IUPAC Name | 4,5,6-trideuteriopyridine-2,3-dicarboxylic acid |
| Molecular Weight (g/mol) | 170.137 |
| Formula Weight | 170.041 g/mol |
| Show More |