Learn More
(S)-(-)-Verapamil-d3 HCl (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | (S)-Verapamil D3 (N-Methyl D3) Hydrochloride |
| CAS | 1398112-33-8 |
| Molecular Formula | C27 D3 H35 N2 O4 . Cl H |
| Quantity | 0.005 g |
| Synonym | (2 S)-2-(3,4-Dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-trideuteriomethylamino]-2-(propan-2-yl)pentanenitrile hydrochloride, (-)-Verapamil-d3 hydrochloride, (S)-Verapamil-d3 hydrochloride, (S)-Verapamil-D3 (N-methyl-D3) hydrochloride, (2 S)-5-[(3,4-Dimethoxyphenethyl)methyl-d3-amino]-2-(3,4-dimethoxyphenyl)-2-isopropylvaleronitrile hydrochloride, (αS)-α-[3-[[2-(3,4-Dimethoxyphenyl)ethyl]methyl-d3-amino]propyl]-3,4-dimethoxy-α-(1-methylethyl)benzeneacetonitrile hydrochloride |
| InChI Formula | InChI=1 S/C27H38N2O4.ClH/c1-20(2)27(19-28,22-10-12-24(31-5)26(18-22)33-7)14-8-15-29(3)16-13-21-9-11-23(30-4)25(17-21)32-6;/h9-12,17-18,20 H,8,13-16H2,1-7H3;1 H/t27-;/m0./s1/i3D3; |
| SMILES | Cl.[2 H]C([2 H])([2 H])N(CCC[C@](C#N)(C(C)C)c1ccc(OC)c(OC)c1)CCc2ccc(OC)c(OC)c2 |
| IUPAC Name | (2 S)-2-(3,4-dimethoxyphenyl)-5-[2-(3,4-dimethoxyphenyl)ethyl-(trideuteriomethyl)amino]-2-propan-2-ylpentanenitrile;hydrochloride |
| Molecular Weight (g/mol) | 494.081 |
| Formula Weight | 493.279 g/mol |
| Show More |