missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-2,2,4,6,6-d5 Heptanoate, CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Specifications
Specifications
| Chemical Name or Material | Testosterone-2,2,4,6,6-D5 17-Heptanoate |
| Molecular Formula | C26 D5 H35 O3 |
| Quantity | 0.005 g |
| Synonym | (17β)-17-(1-Oxoheptyloxy)androst-4-en-3-one-2,2,4,6,6-d5, Testosterone-2,2,4,6,6-d5 heptanoate, Testosterone-2,2,4,6,6-d5 17-heptanoate, Testosterone-2,2,4,6,6-d5 enantate, Testosterone-2,2,4,6,6-d5 17-enanthate, Testosterone-2,2,4,6,6-d5 enanthate |
| InChI Formula | InChI=1 S/C26H40O3/c1-4-5-6-7-8-24(28)29-23-12-11-21-20-10-9-18-17-19(27)13-15-25(18,2)22(20)14-16-26(21,23)3/h17,20-23 H,4-16H2,1-3H3/t20-,21-,22-,23-,25-,26-/m0/s1/i9D2,13D2,17 D |
| SMILES | [2 H]C1=C2C([2 H])([2 H])C[C@H]3 [C@@H]4 CC[C@H](OC(=O)CCCCCC)[C@@]4(C)CC[C@@H]3 [C@@]2(C)CC([2 H])([2 H])C1=O |
| IUPAC Name | [(8 R,9 S,10 R,13 S,14 S,17 S)-2,2,4,6,6-pentadeuterio-10,13-dimethyl-3-oxo-1,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] heptanoate |
| Molecular Weight (g/mol) | 405.625 |
| Formula Weight | 405.329 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction