missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 17beta-Dihydroequilin-2,4,16,16-d4 3-Sulfate (Stabilized with TRIS, 50% w/w), CDN
Click to view available options
Quantity:
0.01 g
0.02 g
Unit Size:
0.01g
0.02g
Specifications
Specifications
| Chemical Name or Material | Sodium 17 beta-Dihydroequilin-2,4,16,16-D4 3-Sulfate |
| CAS | 352431-51-7 |
| Molecular Formula | C18 D4 H17 O5 S . Na |
| Quantity | 0.02 g |
| Synonym | Estra-1,3,5(10),7-tetraene-2,4,16,16-d4-3,17-diol, 3-(hydrogen sulfate), monosodium salt, (17β)- (9 CI), 17β-Dihydroequilin-2,4,16,16-D4 3-sulfate sodium salt, 17β-Dihydroequilin-2,4,16,16-d4 3-sulfate sodium salt, 17β-Dihydroequilin-2,4,16,16-d4 sodium sulfate, (17β)-Estra-1,3,5(10),7-tetraene-3,17-diol-2,4,16,16-d4 3-sulfate sodium salt, Sodium 17β-dihydroequilin-2,4,16,16-D4 3-sulfate |
| InChI Formula | InChI=1 S/C18H22O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h3-5,10,14,16-17,19 H,2,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t14-,16+,17+,18+;/m1./s1/i3D,7D2,10 D; |
| SMILES | [Na+].[2 H]c1cc2[C@H]3 CC[C@@]4(C)[C@@H](CC([2 H])([2 H])[C@@H]4 O)C3=CCc2c([2 H])c1OS(=O)(=O)[O-] |
| IUPAC Name | sodium;[(9 S,13 S,14 S,17 S)-2,4,16,16-tetradeuterio-17-hydroxy-13-methyl-9,11,12,14,15,17-hexahydro-6 H-cyclopenta[a]phenanthren-3-yl] sulfate |
| Molecular Weight (g/mol) | 376.436 |
| Formula Weight | 376.126 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction