Learn More
(±)-Mevalonolactone-4,4,5,5,6,6,6-d7, CDN
Click to view available options
Quantity:
0.025 g
Unit Size:
0.03g
Specifications
Specifications
| Chemical Name or Material | (±)-Mevalolactone-D7 (3-methyl-D3,4,4,5,5-D4) |
| CAS | 347840-19-1 |
| Molecular Formula | C6H3D7O3 |
| Quantity | 0.025 g |
| Synonym | 2 H-Pyran-2-one-5,6-d2, tetrahydro-5,6-d2-4-hydroxy-4-(methyl-d3)- (9 CI), Tetrahydro-4-hydroxy-4-(methyl-d3)-2 H-pyran-2-one-5,5,6,6-d4, (±)-Mevalolactone-d7, (±)-Mevalonolactone-d7, DL-Mevalonic acid lactone-d7, DL-Mevalonic acid delta;-lactone-d7, DL-Mevalonol |
| InChI Formula | InChI=1 S/C6H10O3/c1-6(8)2-3-9-5(7)4-6/h8H,2-4H2,1H3/i1D3,2D2,3D2 |
| SMILES | O=C1OC([2 H])([2 H])C([2 H])([2 H])C(O)(C([2 H])([2 H])[2 H])C1 |
| IUPAC Name | 4-hydroxy-4-(methyl-d3)tetrahydro-2 H-pyran-2-one-5,5,6,6-d4 |
| Molecular Weight (g/mol) | 137.19 |
| Formula Weight | 137.11 g/mol |
| Show More |