Learn More
1-Bromohexadecane-16,16,16-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Bromohexadecane-16,16,16-D3 |
| CAS | 284474-40-4 |
| Molecular Formula | C16 D3 H30 Br |
| Quantity | 0.05 g |
| Synonym | Hexadecane-1,1,1-d3, 16-bromo- (9 CI), 16-Bromohexadecane-1,1,1-d3, 1-Bromohexadecane-16,16,16-d3, 1-Bromohexadecane-16,16,16-D3, 1-Hexadecyl-16,16,16-d3 bromide, Cetyl-16,16,16-d3 bromide |
| InChI Formula | InChI=1 S/C16H33Br/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-16H2,1H3/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])CCCCCCCCCCCCCCCBr |
| IUPAC Name | 16-bromo-1,1,1-trideuteriohexadecane |
| Molecular Weight (g/mol) | 308.3557 |
| Formula Weight | 307.1954 g/mol |
| Show More |