missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Decane-d21-thiol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 1-Decanethiol-D21 |
| CAS | 239090-36-9 |
| Molecular Formula | C10 D21 H S |
| Quantity | 0.05 g |
| Synonym | 1-Decane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-d21-thiol (9 CI), 1-Decane-d21-thiol, 1-Decanethiol-d21, 1-Decanethiol-D21 |
| InChI Formula | InChI=1 S/C10H22S/c1-2-3-4-5-6-7-8-9-10-11/h11H,2-10H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])S |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-henicosadeuteriodecane-1-thiol |
| Molecular Weight (g/mol) | 195.4761 |
| Formula Weight | 195.276 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction