Learn More
2,3,6-Trichloroanisole-d3 (methoxy-d3), CDN
Product Code. 30309380
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309380
Brand: CDN CDND59510.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,3,6-Trichloroanisole D3 (methyl D3)
CAS 1219798-64-7
Molecular Formula C7 D3 H2 Cl3 O
Quantity 0.05 g
Synonym 2,3,6-Trichloroanisole D3 (methyl D3), 2,3,6-Trichloroanisole-d3, 1,2,4-Trichloro-3-(methoxy-d3)benzene
InChI Formula InChI=1 S/C7H5Cl3O/c1-11-7-5(9)3-2-4(8)6(7)10/h2-3 H,1H3/i1D3
SMILES [2 H]C([2 H])([2 H])Oc1c(Cl)ccc(Cl)c1Cl
IUPAC Name 1,2,4-trichloro-3-(trideuteriomethoxy)benzene
Molecular Weight (g/mol) 214.4915
Formula Weight 212.9594 g/mol
Percent Purity 99 atom % D, min 98% Chemical Purity