missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Hydroxyphenyl)ethyl-1,1,2,2-d4-amine HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Tyramine-d4 Hydrochloride |
| CAS | 1189884-47-6 |
| Molecular Formula | C8 2H4 H7 N O . Cl H |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C8H11NO.ClH/c9-6-5-7-1-3-8(10)4-2-7;/h1-4,10 H,5-6,9H2;1 H/i5D2,6D2; |
| SMILES | Cl.[2 H]C([2 H])(N)C([2 H])([2 H])c1ccc(O)cc1 |
| IUPAC Name | 4-(2-amino-1,1,2,2-tetradeuterioethyl)phenol;hydrochloride |
| Molecular Weight (g/mol) | 177.66 |
| Formula Weight | 177.0858 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction