Learn More
4-Chlorobenzyl-2,3,5,6-d4 Cyanide, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | (4-Chlorophenyl)acetonitrile-D4 (4-chlorophenyl-D4) |
| CAS | 1219804-00-8 |
| Molecular Formula | C8 D4 H2 Cl N |
| Quantity | 0.05 g |
| Synonym | Benzene-2,3,5,6-d4-acetonitrile, 4-chloro- (ACI), 4-Chlorobenzene-2,3,5,6-d4-acetonitrile (ACI), 4-Chlorobenzyl-2,3,5,6-d4 Cyanide, (4-Chlorophenyl-d4)acetonitrile, (4-Chlorophenyl)acetonitrile-D4 (4-chlorophenyl-D4) |
| InChI Formula | InChI=1 S/C8H6ClN/c9-8-3-1-7(2-4-8)5-6-10/h1-4 H,5H2/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c(CC#N)c([2 H])c([2 H])c1Cl |
| IUPAC Name | 2-(4-chloro-2,3,5,6-tetradeuteriophenyl)acetonitrile |
| Molecular Weight (g/mol) | 155.6176 |
| Formula Weight | 155.044 g/mol |
| Show More |