missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-4-Hydroxy-3-methoxymandelic-alpha-d1 Acid, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 4-Hydroxy-3-methoxymandelic-alpha-D1 Acid |
| CAS | 53587-34-1 |
| Molecular Formula | C9 D H9 O5 |
| Quantity | 0.05 g |
| Synonym | Benzeneacetic-d acid, α,4-dihydroxy-3-methoxy- (9 CI), Vanillylmandelic-2H2 acid, (±)-4-Hydroxy-3-methoxymandelic-α-d1 Acid, α,4-Dihydroxy-3-methoxybenzeneacetic-d acid, 4-Hydroxy-3-methoxymandelic-d acid, 4-Hydroxy-3-methoxymandelic acid-d1, Vanillylmandelic acid-d1, VMA-D1, 4-Hydroxy-3-methoxymandelic-α-D1 acid |
| InChI Formula | InChI=1 S/C9H10O5/c1-14-7-4-5(2-3-6(7)10)8(11)9(12)13/h2-4,8,10-11 H,1H3,(H,12,13)/i8D |
| SMILES | [2 H]C(O)(C(=O)O)c1ccc(O)c(OC)c1 |
| IUPAC Name | 2-deuterio-2-hydroxy-2-(4-hydroxy-3-methoxyphenyl)acetic acid |
| Molecular Weight (g/mol) | 199.1789 |
| Formula Weight | 199.0591 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction