Learn More
Anthranilic-3,4,5,6-d4 Acid, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Anthranilic-3,4,5,6-d4 acid |
| CAS | 60124-83-6 |
| Molecular Formula | C7 2H4 H3 N O2 |
| Quantity | 0.05 g |
| Synonym | Benzoic-2,3,4,5-d4 acid, 6-amino-, 6-Aminobenzoic-2,3,4,5-d4 acid, 2-Aminobenzoic-3,4,5,6-d4 acid, Anthranilic acid-d4, Anthranilic acid-d4 (phenyl-d4), Anthranilic-3,4,5,6-D4 acid |
| InChI Formula | InChI=1 S/C7H7NO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4 H,8H2,(H,9,10)/i1D,2 D,3 D,4 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)O)c(N)c1[2 H] |
| IUPAC Name | 2-amino-3,4,5,6-tetradeuteriobenzoic acid |
| Molecular Weight (g/mol) | 141.16 |
| Formula Weight | 141.0728 g/mol |
| Show More |