missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-Atenolol-d7 (iso-propyl-d7), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Atenolol-d7 |
| CAS | 1202864-50-3 |
| Molecular Formula | C14 D7 H15 N2 O3 |
| Quantity | 0.05 g |
| Synonym | Atenolol D7, Benzeneacetamide, 4-[2-hydroxy-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]propoxy]-, 4-[2-Hydroxy-3-[[1-(methyl-d3)ethyl-1,2,2,2-d4]amino]propoxy]benzeneacetamide, Atenolol-d7, Atenolol D7 (Isopropyl D7) |
| InChI Formula | InChI=1 S/C14H22N2O3/c1-10(2)16-8-12(17)9-19-13-5-3-11(4-6-13)7-14(15)18/h3-6,10,12,16-17 H,7-9H2,1-2H3,(H2,15,18)/i1D3,2D3,10 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NCC(O)COc1ccc(CC(=O)N)cc1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-[4-[3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)-2-hydroxypropoxy]phenyl]acetamide |
| Molecular Weight (g/mol) | 273.379 |
| Formula Weight | 273.207 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction