missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(6-methylheptyl) Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic Acid Bis-6-methylheptyl Ester-D4 (phthalic-D4) |
| CAS | 2209087-06-7 |
| Molecular Formula | C24 D4 H34 O4 |
| Quantity | 0.05 g |
| Synonym | Bis(6-methylheptyl) Phthalate-3,4,5,6-d4, Phthalic Acid Bis-6-methylheptyl Ester-d4, Di(6-methylheptyl) phthalate-d4, Diisooctyl phthalate-d4, 1,2-Benzenedicarboxylic-3,4,5,6-d4 acid 1,2-bis(6-methylheptyl) ester, Phthalic Acid Bis-6-methylheptyl Ester-D4 (phthalic-D4) |
| InChI Formula | InChI=1 S/C24H38O4/c1-19(2)13-7-5-11-17-27-23(25)21-15-9-10-16-22(21)24(26)28-18-12-6-8-14-20(3)4/h9-10,15-16,19-20 H,5-8,11-14,17-18H2,1-4H3/i9D,10 D,15 D,16 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OCCCCCC(C)C)c(C(=O)OCCCCCC(C)C)c1[2 H] |
| IUPAC Name | bis(6-methylheptyl) 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 394.5808 |
| Formula Weight | 394.3021 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction