Learn More
Dimethyl Phthalate-3,4,5,6-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | Phthalic acid, bis-methyl ester D4 |
| CAS | 93951-89-4 |
| Molecular Formula | C10 2H4 H6 O4 |
| Quantity | 0.05 g |
| Synonym | 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, 1,2-dimethyl ester, 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, dimethyl ester (9 CI), Phthalic acid dimethyl ester D4 |
| InChI Formula | InChI=1 S/C10H10O4/c1-13-9(11)7-5-3-4-6-8(7)10(12)14-2/h3-6 H,1-2H3/i3D,4 D,5 D,6 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC)c(C(=O)OC)c1[2 H] |
| IUPAC Name | dimethyl 3,4,5,6-tetradeuteriobenzene-1,2-dicarboxylate |
| Molecular Weight (g/mol) | 198.21 |
| Formula Weight | 198.083 g/mol |
| Show More |