missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hexafluorobisphenol A-3,3',5,5'-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2,2-Bis-(4-hydroxyphenyl)hexafluoropropane-d4 |
| Molecular Formula | C15 D4 H6 F6 O2 |
| Quantity | 0.05 g |
| Synonym | Hexafluorobisphenol A-3,3',5,5'-d4, Bisphenol AF-3,3',5,5'-D4, 4,4'-[2,2,2-Trifluoro-1-(trifluoromethyl)ethylidene]bis[phenol-2,6-d2], 2,2-Bis(4-hydroxyphenyl-3,5-d2)-1,1,1,3,3,3-hexafluoropropane |
| InChI Formula | InChI=1 S/C15H10F6O2/c16-14(17,18)13(15(19,20)21,9-1-5-11(22)6-2-9)10-3-7-12(23)8-4-10/h1-8,22-23 H/i5D,6 D,7 D,8 D |
| SMILES | [2 H]c1cc(cc([2 H])c1O)C(c2cc([2 H])c(O)c([2 H])c2)(C(F)(F)F)C(F)(F)F |
| IUPAC Name | 2,6-dideuterio-4-[2-(3,5-dideuterio-4-hydroxyphenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl]phenol |
| Molecular Weight (g/mol) | 340.254 |
| Formula Weight | 340.084 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction