missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isofenphos-d7 (N-iso-propyl-d7), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Isofenphos-D7 (N-isopropyl-D7) |
| Molecular Formula | C15 D7 H17 N O4 P S |
| Quantity | 0.05 g |
| Synonym | Isofenphos-D7 (N-isopropyl-D7), Isofenphos-d7, 1-(Methyl-d3)ethyl-1,2,2,2-d4 2-[[ethoxy[(1-methylethyl)amino]phosphinothioyl]oxy]benzoate, 2-[[Ethoxy[(1-methylethyl)amino]phosphinothioyl]oxy]benzoic acid 1-(methyl-d3)ethyl-1,2,2,2-d4 ester |
| InChI Formula | InChI=1 S/C15H24NO4PS/c1-6-18-21(22,16-11(2)3)20-14-10-8-7-9-13(14)15(17)19-12(4)5/h7-12 H,6H2,1-5H3,(H,16,22)/i2D3,3D3,11 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(NP(=S)(OCC)Oc1ccccc1C(=O)OC(C)C)C([2 H])([2 H])[2 H] |
| IUPAC Name | propan-2-yl 2-[ethoxy-(1,1,1,2,3,3,3-heptadeuteriopropan-2-ylamino)phosphinothioyl]oxybenzoate |
| Molecular Weight (g/mol) | 352.4373 |
| Formula Weight | 352.1603 g/mol |
| Percent Purity | 99 atom % D, min 95% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction