Learn More
L-4-Hydroxyphenyl-2,6-d2-alanine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | L-Tyrosine-D2 (4-hydroxyphenyl-2,6-D2) |
| CAS | 57746-15-3 |
| Molecular Formula | C9 D2 H9 N O3 |
| Quantity | 0.05 g |
| Synonym | L-Tyrosine-2,6-d2, L-Tyrosine-2',6'-2H2, L-(2,6-2H2)Tyrosine, L-2,6-2H2-Tyrosine, L-Tyrosine-2H2, (2 S)-2-Amino-3-(4-hydroxyphenyl-2,6-d2)propanoic acid, (2 S)-2-Amino-3-(4-hydroxyphenyl-2,6-d2)propanoic acid, (2 S)-2-Amino-3-(4-hydroxyphenyl-2,6-d2)propanoic acid, (2 S)-2-Amino-3-(4-hydroxyphenyl-2,6-d2)propanoic acid, (2 S)-2-Amino-3-(4-hydroxyphenyl-2,6-d2)propanoic acid |
| InChI Formula | InChI=1 S/C9H11NO3/c10-8(9(12)13)5-6-1-3-7(11)4-2-6/h1-4,8,11 H,5,10H2,(H,12,13)/t8-/m0/s1/i1D,2 D |
| SMILES | [2 H]c1cc(O)cc([2 H])c1C[C@H](N)C(=O)O |
| IUPAC Name | (2 S)-2-amino-3-(2,6-dideuterio-4-hydroxyphenyl)propanoic acid |
| Molecular Weight (g/mol) | 183.201 |
| Formula Weight | 183.086 g/mol |
| Show More |