missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(6R,12aR)-Tadalafil-d3 (N-methyl-d3), CDN
Click to view available options
Quantity:
0.005 g
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | Tadalafil-d3 |
| CAS | 960226-55-5 |
| Molecular Formula | C22 2H3 H16 N3 O4 |
| Quantity | 0.01 g |
| InChI Formula | InChI=1 S/C22H19N3O4/c1-24-10-19(26)25-16(22(24)27)9-14-13-4-2-3-5-15(13)23-20(14)21(25)12-6-7-17-18(8-12)29-11-28-17/h2-8,16,21,23 H,9-11H2,1H3/t16-,21-/m1/s1/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])N1CC(=O)N2[C@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O |
| IUPAC Name | (2 R,8 R)-2-(1,3-benzodioxol-5-yl)-6-(trideuteriomethyl)-3,6,17-triazatetracyclo[8.7.0.0 ^{3,8}0.0 ^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione |
| Molecular Weight (g/mol) | 392.42 |
| Formula Weight | 392.1564 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction