missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mepiquat-d16 Chloride, CDN
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Specifications
Specifications
| Chemical Name or Material | Mepiquat-d16 Chloride |
| Molecular Formula | C7 D16 N . Cl |
| Quantity | 0.05 g |
| Synonym | 1,1-Di(methyl-d3)piperidin-2,2,3,3,4,4,5,5,6,6-d10-ium chloride, Mepiquat-d16 chloride, N,N-Dimethylpiperidinium-d16 chloride, Mepiquat-D16 chloride |
| InChI Formula | InChI=1 S/C7H16N.ClH/c1-8(2)6-4-3-5-7-8;/h3-7H2,1-2H3;1 H/q+1;/p-1/i1D3,2D3,3D2,4D2,5D2,6D2,7D2; |
| SMILES | [Cl-].[2 H]C([2 H])([2 H])[N+]1(C([2 H])([2 H])[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C1([2 H])[2 H] |
| IUPAC Name | 2,2,3,3,4,4,5,5,6,6-decadeuterio-1,1-bis(trideuteriomethyl)piperidin-1-ium;chloride |
| Molecular Weight (g/mol) | 165.7602 |
| Formula Weight | 165.1976 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction