Learn More
Methyl Viologen-d8 Dichloride (rings-d8), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Paraquat dichloride D8 |
| CAS | 347841-45-6 |
| Molecular Formula | C12 2H8 H6 N2 . 2 Cl |
| Quantity | 0.05 g |
| InChI Formula | InChI=1 S/C12H14N2.2 ClH/c1-13-7-3-11(4-8-13)12-5-9-14(2)10-6-12;;/h3-10 H,1-2H3;2*1 H/q+2;;/p-2/i3D,4 D,5 D,6 D,7 D,8 D,9 D,10 D;; |
| SMILES | [Cl-].[Cl-].[2 H]c1c([2 H])[n+](C)c([2 H])c([2 H])c1c2c([2 H])c([2 H])[n+](C)c([2 H])c2[2 H] |
| IUPAC Name | 2,3,5,6-tetradeuterio-1-methyl-4-(2,3,5,6-tetradeuterio-1-methylpyridin-1-ium-4-yl)pyridin-1-ium;dichloride |
| Molecular Weight (g/mol) | 265.21 |
| Formula Weight | 264.1036 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |