Learn More
N-(2-Methyl-d3-benzoyl-d4)glycine, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Methyl Hippuric Acid-d7 |
| CAS | 1216430-90-8 |
| Molecular Formula | C10 D7 H4 N O3 |
| Quantity | 0.05 g |
| Synonym | 2-[[2,3,4,5-Tetradeuterio-6-(trideuteriomethyl)benzoyl]amino]acetic acid, N-(2-Methylbenzoyl)glycine-D7 (methylbenzoyl-D7), N-(2-Methylbenzoyl)glycine-d7, 2-Methylhippuric acid-d7, 2-[[2-(Methyl-d3)benzoyl-3,4,5,6-d4]amino]acetic acid, o-Methylhippuric acid-d7, o-Toluric acid-d7, 2-Methyl hippuric acid-d7 |
| InChI Formula | InChI=1 S/C10H11NO3/c1-7-4-2-3-5-8(7)10(14)11-6-9(12)13/h2-5 H,6H2,1H3,(H,11,14)(H,12,13)/i1D3,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c(C(=O)NCC(=O)O)c1[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-[[2,3,4,5-tetradeuterio-6-(trideuteriomethyl)benzoyl]amino]acetic acid |
| Molecular Weight (g/mol) | 200.242 |
| Formula Weight | 200.118 g/mol |
| Show More |