missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Hydroxyphenyl)acetamide-2,2,2-d3, CDN
Click to view available options
Quantity:
0.05 g
0.25 g
Unit Size:
0.05g
0.25g
Specifications
Specifications
| Chemical Name or Material | Acetaminophen methyl-d3 |
| CAS | 60902-28-5 |
| Molecular Formula | C8 2H3 H6 N O2 |
| Quantity | 0.05 g |
| Synonym | Acetaminophen D3 (Methyl D3), Acetamide-2,2,2-d3, N-(4-hydroxyphenyl)-, N-(4-Hydroxyphenyl)acetamide-2,2,2-d3, Paracetamol D3 |
| InChI Formula | InChI=1 S/C8H9NO2/c1-6(10)9-7-2-4-8(11)5-3-7/h2-5,11 H,1H3,(H,9,10)/i1D3 |
| SMILES | [2 H]C([2 H])([2 H])C(=O)Nc1ccc(O)cc1 |
| IUPAC Name | 2,2,2-trideuterio-N-(4-hydroxyphenyl)acetamide |
| Molecular Weight (g/mol) | 154.18 |
| Formula Weight | 154.0822 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction