Learn More
n-Undecyl-d23-amine, CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | 1-Undecanamine-D23 |
| CAS | 1219805-57-8 |
| Molecular Formula | C11 D23 H2 N |
| Quantity | 0.05 g |
| Synonym | 1-Undecan-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-d23-amine, Undecanamine-d23, n-Undecyl-d23 amine, 1-Undecanamine-d23, 1-Undecanamine-D23, 1-Aminoundecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-d23 |
| InChI Formula | InChI=1 S/C11H25N/c1-2-3-4-5-6-7-8-9-10-11-12/h2-12H2,1H3/i1D3,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,11-tricosadeuterioundecan-1-amine |
| Molecular Weight (g/mol) | 194.465 |
| Formula Weight | 194.343 g/mol |
| Show More |