missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Butane-d8-diamine, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine |
| CAS | 709608-92-4 |
| Molecular Formula | C4 2H8 H4 N2 |
| Quantity | 0.1 g |
| Synonym | 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine (9 CI), 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine, 1,4-Butane-d8-diamine, 1,4-Butanediamine-(1,1,2,2,3,3,4,4-D8), 1,1,2,2,3,3,4,4-Octadeuteriobutane-1,4-diamine |
| InChI Formula | InChI=1 S/C4H12N2/c5-3-1-2-4-6/h1-6H2/i1D2,2D2,3D2,4D2 |
| SMILES | [2 H]C([2 H])(N)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2,3,3,4,4-octadeuteriobutane-1,4-diamine |
| Molecular Weight (g/mol) | 96.2 |
| Formula Weight | 96.1503 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction