Learn More
2,4-Dihydroxybenzophenone-2',3',4',5',6'-d5, CDN
Product Code. 30321857
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.10g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30321857
Brand: CDN CDND76450.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2,4-Dihydroxybenzophenone-d5
CAS 91586-06-0
Molecular Formula C13 D5 H5 O3
Quantity 0.1 g
Synonym 2,4-Dihydroxybenzophenone D5 (Phenyl D5)
InChI Formula InChI=1 S/C13H10O3/c14-10-6-7-11(12(15)8-10)13(16)9-4-2-1-3-5-9/h1-8,14-15 H/i1D,2 D,3 D,4 D,5 D
SMILES [2 H]c1c([2 H])c([2 H])c(C(=O)c2ccc(O)cc2O)c([2 H])c1[2 H]
IUPAC Name (2,4-dihydroxyphenyl)-(2,3,4,5,6-pentadeuteriophenyl)methanone
Molecular Weight (g/mol) 219.248
Formula Weight 219.094 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity