missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-4-chlorophenol-d6, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Amino-4-chlorophenol-D6 |
| Molecular Formula | C6 D6 Cl N O |
| Quantity | 0.1 g |
| Synonym | 2-Amino-4-chlorophenol-N,N,O,3,5,6-d6, 2-Amino-4-chlorophenol-D6, 2-Amino-4-chlorophenol-d6, 5-Chloro-2-hydroxyaniline-d6 |
| InChI Formula | InChI=1 S/C6H6ClNO/c7-4-1-2-6(9)5(8)3-4/h1-3,9 H,8H2/i1D,2 D,3 D/hD3 |
| SMILES | [2 H]Oc1c([2 H])c([2 H])c(Cl)c([2 H])c1N([2 H])[2 H] |
| IUPAC Name | 3-chloro-N,N,2,4,5-pentadeuterio-6-deuteriooxyaniline |
| Molecular Weight (g/mol) | 149.6079 |
| Formula Weight | 149.0515 g/mol |
| Percent Purity | 98 atom % D, min 97% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction