Learn More
2-Butanone-d8, CDN
Product Code. 30317763
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30317763
Brand: CDN CDND43310.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 2-Butanone-D8
CAS 350820-09-6
Molecular Formula C4 D8 O
Quantity 0.1 g
Synonym 2-Butanone-1,1,1,3,3,4,4,4-d8 (9 CI), 2-Butanone-d8, 2-Butanone-D8, Methyl-d3 ethyl-d5 ketone
InChI Formula InChI=1 S/C4H8O/c1-3-4(2)5/h3H2,1-2H3/i1D3,2D3,3D2
SMILES [2 H]C([2 H])([2 H])C(=O)C([2 H])([2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,1,1,3,3,4,4,4-octadeuteriobutan-2-one
Molecular Weight (g/mol) 80.155
Formula Weight 80.1077 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity