missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2RS,5R)-Menthone-2,6,6-d3, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | (2 RS,5 R)-Menthone-2,6,6-D3 |
| Molecular Formula | C10 D3 H15 O |
| Quantity | 0.1 g |
| Synonym | (2 RS,5 R)-Menthone-2,6,6-d3, (2 RS,5 R)-5-Methyl-2-(1-methylethyl)cyclohexanone-2,2,6-d3 |
| InChI Formula | InChI=1 S/C10H18O/c1-7(2)9-5-4-8(3)6-10(9)11/h7-9 H,4-6H2,1-3H3/t8-,9?/m1/s1/i6D2,9 D |
| SMILES | [2 H]C1([2 H])[C@H](C)CCC([2 H])(C(C)C)C1=O |
| IUPAC Name | (3 R)-2,2,6-trideuterio-3-methyl-6-propan-2-ylcyclohexan-1-one |
| Molecular Weight (g/mol) | 157.2678 |
| Formula Weight | 157.1546 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction