missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(±)-3-Methyl-d3-3-hexanol-2,2,4,4-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 3-Methyl-3-hexanol-1',1',1',2,2,4,4-D7 |
| Molecular Formula | C7 D7 H9 O |
| Quantity | 0.1 g |
| Synonym | (±)-3-Methyl-d3-3-hexanol-2,2,4,4-d4, 3-Methyl-3-hexanol-d7, 3-Methyl-3-hexanol-1',1',1',2,2,4,4-D7, 2-(Ethyl-1,1-d2)-2-pentanol-1,1,1,3,3-d5 |
| InChI Formula | InChI=1 S/C7H16O/c1-4-6-7(3,8)5-2/h8H,4-6H2,1-3H3/i3D3,5D2,6D2 |
| SMILES | [2 H]C([2 H])([2 H])C(O)(C([2 H])([2 H])C)C([2 H])([2 H])CC |
| IUPAC Name | 2,2,4,4-tetradeuterio-3-(trideuteriomethyl)hexan-3-ol |
| Molecular Weight (g/mol) | 123.2445 |
| Formula Weight | 123.1641 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction